Please login to your online account to display your discounted pricing

eBioscience™ Mouse TGF beta 1, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

$1,825.55 - $6,088.55

Specifications

Structural Form TGFb1 was expressed in CHO cells.
Mature TGFb1 (ala279 - ser390; accession #BC001180) was
purified from the conditioned media.
Purity >95%
Purification Method SDS-PAGE
Molecular Weight Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25kDa protein.
Concentration 0.5mg/mL
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123502

 
ebioscience™
34834285
500μg Each for $6,088.55

501129280

 
ebioscience™
34834282
100μg Each for $1,825.55
Description & Specifications

Specifications

Structural Form TGFb1 was expressed in CHO cells.
Mature TGFb1 (ala279 - ser390; accession #BC001180) was
purified from the conditioned media.
Purity >95%
Purification Method SDS-PAGE
Molecular Weight Mature TGFb1 polypeptide has a predicted molecular mass of 12,794. On reducing SDS-PAGE the protein migrates as a 13kDa polypeptide and on non-reducing SDS page the cystine-linked homodimer migrates as a 25kDa protein.
Concentration 0.5mg/mL
Source CHO Cells
Accession Number BC001180
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Formulation Sterile liquid; 0.1M Gly-HCl, pH 3.0. 0.22 µm
filtered.
Biological Activity The ED50 of this protein, as measured by inhibition of IL-4 induced proliferation in HT-2 cells, is less than or equal to 40pg/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 107 units/mg.
For Use With (Application) Functional Assays, Cell Culture
Validated Application FA
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Mouse
Gene Symbol Transforming growth factor beta-1
Name TGF beta 1
Recombinant Yes

Mouse TGF beta 1 (Transforming Growth Factor beta 1) is a member of a superfamily of homologous, disulfide-linked, homodimeric proteins that regulate the proliferation and differentiation of normal and transformed cells. Human TGF beta 1 is a 25 kDa protein with each subunit containing 112 aa residues. TGF beta 1 is secreted from the many cell types in a complex with LAP (Latency Associated Peptide) and also with LTBP (latent TGF beta binding protein). Disassociation is thought to happen in the extracellular matrix via proteases and/or integrins. Regulatory T cells are a recently identified source of TGF beta 1 thought to be a key player in their suppressive functions. TGF beta 1 binds through TGF beta receptors (of which there is a large family). The protein is highly conserved across all mammalian species.