Please login to your online account to display your discounted pricing

eBioscience™ Mouse Stem Cell Factor (SCF), Recombinant Protein

Mouse Recombinant Protein

$201.55 - $317.55

Specifications

Name Stem Cell Factor (SCF)
Recombinant Yes
Conjugate Unlabeled
Purity ≥95%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018498

 
life technologies
LS14834162
5μg Each for $201.55

5018499

 
life technologies
LS14834163
10μg Each for $317.55
Description & Specifications

Specifications

Name Stem Cell Factor (SCF)
Recombinant Yes
Conjugate Unlabeled
Purity ≥95%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight The protein is not methionylated at the N-terminal and has a predicted molecular mass of 18,298. The DTT reduced protein migrates as an 18kDa polypeptide on SDS-PAGE. The non-reduced protein migrates as a 16kDa polypeptide.
Storage Requirements Store at less than or equal to -70°C.

Mouse Stem Cell Factor (SCF) is a hematopoietic growth factor and ligand for the KIT tyrosine kinase receptor, which exerts its activity in the early stages of hematopoiesis. Mouse SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures.