missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Mouse ST2 (IL-33R) ELISA Ready-SET-Go!™ Kit
Specifically engineered for accurate and precise measurement of mouse ST2 (IL-33R) protein levels from samples including serum and cell culture supernatants.
Manufacturer: Invitrogen™ LS88933476
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Sample Diluent B
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.
Description
Mouse ST2 (IL-33R) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
ELISA | |
20 x 96 tests | |
Mouse | |
IL-33R (ST2) |
96-well plates | |
80pg/mL | |
2° to 8°C except standard which should be stored at ≤−70°C | |
IL1RL1, Interleukin-1 Receptor-Like 1, Ly84 |
For Research Use Only