Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse Soluble ICAM-1 (CD54) ELISA Kits


Measure levels of mouse soluble ICAM-1 (CD54) in mouse serum, plasma (EDTA) and cell culture supernatant samples using 96-well plates.

Manufacturer: thermo scientific™ pierce™  EMICAM12

 View more versions of this product

Catalog No. ENEMICAM12

  • / Each
 

Description & Specifications

Specifications

Precision Intra-assay CV <10%; inter assay CV <10%
Sensitivity <5pg/mL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Formulation Complete ELISA kit with coated 96-well plate
Kit Contents 30X Wash Buffer, 2 x 50mL
Assay Range 25.6 to 1000pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 2 plate kit
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50µL of 100-fold diluted samples per well
Specificity No significant cross reaction with rat or human sICAM 1
Total Assay Time 3.5 hr.
Target Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)
Description Mouse Soluble ICAM-1 (CD54) ELISA Kit
Sufficient For 96 microplate wells
Item Description 2 x 96 microplate wells

Assay sICAM-1 (mouse soluble ICAM-1, also called CD54) levels in mouse serum, plasma (EDTA) and culture supernatant samples with our Thermo Scientific Mouse Soluble ICAM (CD54) ELISA Kits.

These mouse sICAM-1 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54) in samples at 50μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Mouse soluble ICAM-1 (intercellular adhesion molecule 1, also called CD54)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 25.6 to 1000ng/mL

  • Sensitivity: < 5ng/mL

  • Precision: Intra-assay CV < 10%; inter assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL of 100 fold diluted samples per well

  • Specificity: No significant cross reaction with rat or human sICAM 1

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.