Please login to your online account to display your discounted pricing

eBioscience™ Mouse sCD40L Platinum ELISA Kit

For the quantitative detection of mouse sCD40L.

$651.05 - $3,116.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym TRAP, HIGM 1, T cell antigen Gp39, CD154
Target sCD154 (sCD40 Ligand)
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse sCD40L
Biotin-Conjugate anti-mouse sCD40L monoclonal antibody
Streptavidin-HRP
Mouse sCD40L Standard lyophilized, 40ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125440

 
life technologies
LSBMS6010
96 tests Each for $651.05

501125441

 
life technologies
LSBMS6010TEN
10 x 96 tests Each for $3,116.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym TRAP, HIGM 1, T cell antigen Gp39, CD154
Target sCD154 (sCD40 Ligand)
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 0.14ng/mL
Species Mouse

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.