missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse sCD40L Platinum ELISA Kit

For the quantitative detection of mouse sCD40L.

Manufacturer:  Invitrogen™ LSBMS6010TEN

 View more versions of this product

Catalog No. 501125441



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse sCD40L
Biotin-Conjugate anti-mouse sCD40L monoclonal antibody
Streptavidin-HRP
Mouse sCD40L Standard lyophilized, 40ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum
Mouse
TRAP, HIGM 1, T cell antigen Gp39, CD154
ELISA
0.14ng/mL
50μL
sCD154 (sCD40 Ligand)
Documents

For Research Use Only