Please login to your online account to display your discounted pricing

Mouse/Rat Total Cytochrome c DuoSet IC, Short Format, R&D Systems


Manufacturer: r & d systems  RDSDYC8975

 View more versions of this product

Catalog No. DYC8975

  • / Each
 

Description & Specifications

Specifications

Species Mouse, rat
Product Type ELISA kit
Format 5 x 96-well plate
Quantity 1 kit
Applications Development of sandwich ELISA to measure cytochrome c in cell lysates