Please login to your online account to display your discounted pricing

Mouse/Rat Total Cytochrome c DuoSet IC, Short Format, R&D Systems


Manufacturer: r & d systems  RDSDYC8972

 View more versions of this product

Catalog No. DYC8972

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Format 2 x 96-well plate
Product Type ELISA kit
Quantity 1 kit
Species Mouse, rat