missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse/Rat Total Cytochrome c DuoSet IC, Short Format, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

$428.87 - $846.01

Specifications

Applications ELISA
Product Type ELISA kit
Quantity 1 kit
Species Mouse, rat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
DYC8972 R & D Systems
RDSDYC8972
2 x 96-well plate Each for $428.87
DYC8975 R & D Systems
RDSDYC8975
5 x 96-well plate Each for $846.01
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

ELISA
ELISA kit
1 kit
Mouse, rat