Please login to your online account to display your discounted pricing

Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA Kit, R&D Systems

$663.51 - $3,180.69

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Heparin Plasma (40μL)
Species Mouse, Rat, Porcine, Canine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MB100B

 
r & d systems
RDSMB100B
1 Plate Each for $663.51

SMB100B

 
r & d systems
RDSSMB100B
6 Plates Each for $3,180.69
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Heparin Plasma (40μL)
Species Mouse, Rat, Porcine, Canine

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Heparin Plasma (40μL)
Sensitivity: 15.4pg/mL
Specificity: Natural and recombinant TGF-beta 1. This kit also recognizes human TGF-beta 1.2.
Cross Reactivity: Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.