Please login to your online account to display your discounted pricing

Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSMB100B

 View more versions of this product

Catalog No. SMB100B

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 6 Plates
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Species Mouse, Rat, Porcine, Canine
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type Solid Phase Sandwich ELISA
Sensitivity 15.4pg/mL
Gene ID (Entrez) 21803
Sample Volume Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Heparin Plasma (40μL)
Quantity 1 Pk.