Please login to your online account to display your discounted pricing

Mouse/Rat Leptin Quantikine ELISA Kit, R&D Systems

$802.62 - $1,925.51

Specifications

Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
Sensitivity 22pg/mL
Species Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Gene ID (Entrez) Quantity Price Quantity    

MOB00

 
r & d systems
RDSMOB00
25608 1 Kit Each for $802.62

SMOB00

 
r & d systems
RDSSMOB00
16846 1 Pk. Each for $1,925.51
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
Sensitivity 22pg/mL
Species Mouse, Rat

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL)
Sensitivity: 22pg/mL
Specificity: Natural and recombinant mouse and rat Leptin
Cross Reactivity: Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules.