Please login to your online account to display your discounted pricing

Mouse/Rat Leptin Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSMOB00

 View more versions of this product

Catalog No. SMOB00

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 16846
Quantity 1 Pk.
Sensitivity 22pg/mL
Sample Type Cell Culture Supernates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL)
Species Mouse, Rat
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates