Please login to your online account to display your discounted pricing

Mouse/Rat IGF-I Quantikine ELISA Kit, R&D Systems

$611.21 - $2,933.57

Specifications

Sufficient For Mouse, Rat
View More Specs
Products
Catalog Number Mfr. No. Gene ID (Entrez) Product Type Quantity Price Quantity    

MG100

 
r & d systems
RDSMG100
16000 IGF-I Quantikine ELISA Kit 1 kit Each for $611.21

SMG100

 
r & d systems
RDSSMG100
24482 IGF-I Quantikine ELISA Kit 1 Pk. Each for $2,933.57
Description & Specifications

Specifications

Sufficient For Mouse, Rat

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (25μL), Tissue Homogenates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity: 8.4pg/mL
Specificity: Natural and recombinant mouse and rat IGF-I
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.