Please login to your online account to display your discounted pricing

Mouse/Rat CCL2/JE/MCP-1 Quantikine ELISA Kit, R&D Systems


A 4.5 hour solid phase ELISA designed to measure MCP-1 in mouse or rat cell culture supernates and serum. It contains E. coli-expressed mouse MCP-1 and antibodies raised against the recombinant factor.

Manufacturer: r & d systems  RDSMJE00

 View more versions of this product

Catalog No. MJE00

  • / Each
 

Description & Specifications

Specifications

Sample Type Cell Culture Supernates, Serum
Sufficient For Sufficient materials to run ELISAs on at least two 96-well plates
Quantity 1 kit
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Gene ID (Entrez) 20296 (Mouse); 24770 (Rat)
Sensitivity 2pg/mL
Species Mouse, Rat
Sample Volume Cell Culture Supernates (25μL), Serum (25μL)

MCP-1, also known as monocyte chemotactic and activating factor (MCAF), was initially purified based on its ability to chemoattract monocytes. Subsequent to its cloning and sequencing, it became evident that this protein is also identical to the product of the human JE gene. The JE gene, originally identified in mouse fibroblasts, is a platelet-derived growth factor (PDGF)-inducible gene.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (25μL), Serum (25μL)
Sensitivity: 2pg/mL
Specificity: Natural and recombinant mouse and rat MCP-1
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.