Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse MIP-1 alpha (CCL3) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of mouse MIP-1 alpha protein levels from samples including serum, plasma and cell culture supernatants. MIP-1 alpha and MIP-1 beta belong to the family of cysteine-cysteine (cc) chemokines.

$361.05 - $651.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Small inducible cytokine A3, SCYA3, CCL3
Target CCL3 (MIP-1 alpha)
Sample Type Cell culture supernatant, serum and plasma
View More Specs
Includes:  Capture Antibody Concentrate
Detection Antibody Concentrate
Mouse MIP-1 alpha Standard (lyophilized): 100pg/mL upon reconstitution
Coating Buffer Concentrate
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
pre-titrated Streptavidin-HRP
Tetramethylbenzidine (TMB) Substrate Solution
96 Well Plate included with product Cat. #'s ending in suffixes -22, -76, -86
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125171

 
ebioscience
LS885601322
96-well plates 2 x 96 tests Each for $361.05

501125172

 
ebioscience
LS885601388
Each for $651.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Small inducible cytokine A3, SCYA3, CCL3
Target CCL3 (MIP-1 alpha)
Sample Type Cell culture supernatant, serum and plasma
Sensitivity 0.8pg/mL
Species Mouse

Mouse MIP-1 alpha (CCL3) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)