Please login to your online account to display your discounted pricing

eBioscience™ Mouse MCPT-1 (mMCP-1) ELISA Ready-SET-Go!™ Kit

Optimized for the accurate and precise measurement of mouse MCPT-1 in serum, and tissue culture supernatant samples. MCPT-1 (Mast Cell Protease-1) is a β-chymase, a type of serine protease stored and secreted in a tissue-specific manner by mucosal mast cells.

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym Mouse Mast Cell Protease-1
Target MCPT-1 (mMCP-1)
Sensitivity 120pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017416

 
life technologies
LS88750322
Each for $332.05

5017417

 
life technologies
LS88750386
96-well plates 10 x 96 tests Each for $694.55

5017418

 
life technologies
LS88750388
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Mouse Mast Cell Protease-1
Target MCPT-1 (mMCP-1)
Sensitivity 120pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Mouse

Mouse MCPT-1 Ready-SET-Go!™ reagent set includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays