missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ IL-9 Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement of IL-9 protein levels from samples including serum, and cell culture supernatants. IL-9 is a 14 kd cytokine originally named P40 and identified by its proliferative effects on T cell population
$390.00 - $1,240.00
Specifications
Target Species | Mouse |
---|---|
For Use With (Application) | ELISA |
Synonym | Interleukin-9, T-cell growth factor P40 |
Catalog Number | Mfr. No. | Quantity | Gene Symbol | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Gene Symbol | Price | Quantity | ||||
501125213
|
Invitrogen™
88809222 |
2 x 96 Tests | CED,DPD1,LAP,TGFB,TGFbeta |
N/A
|
|
||||
501125217
|
Invitrogen™
88809288 |
10 x 96 Tests | IL-2 |
N/A
|
|
||||
501125215
|
Invitrogen™
88809277 |
20 x 96 Tests | IL-2 |
N/A
|
|
||||
Description
Mouse IL-9 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Mouse | |
ELISA | |
Interleukin-9, T-cell growth factor P40 |
For Research Use Only