Invitrogen™ eBioscience™ Mouse IL-9 ELISA Ready-SET-Go!™ Kit, 2nd Generation
Specifically engineered for accurate and precise measurement of IL-9 protein levels from samples including serum, and cell culture supernatants. IL-9 is a 14 kd cytokine originally named P40 and identified by its proliferative effects on T cell population
Manufacturer:  Invitrogen™ LS88809276
 View more versions of this product
Catalog No. 501125214
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.
Description
Description
Mouse IL-9 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
20 x 96 tests
Mouse
IL-9
96-well plates
32pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-9, T-cell growth factor P40