Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-7, Recombinant Protein

Mouse Recombinant Protein

$303.05 - $303.05

Specifications

Structural Form E. coli expressed amino acids glu 26-ile 154 of mature mouse IL-7 accession # NM_008371
Purity >98%
Purification Method SDS-PAGE
Molecular Weight The methionylated polypeptide has a predicted molecular mass of 15,028. The DTT reduced protein migrates as a 15kDa polypeptide. The non-reduced protein has slightly higher mobility on non-reducing SDS-PAGE.
Source E. coli
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501128868

 
ebioscience™
LS14807162
5μg Each for $303.05
Description & Specifications

Specifications

Structural Form E. coli expressed amino acids glu 26-ile 154 of mature mouse IL-7 accession # NM_008371
Purity >98%
Purification Method SDS-PAGE
Molecular Weight The methionylated polypeptide has a predicted molecular mass of 15,028. The DTT reduced protein migrates as a 15kDa polypeptide. The non-reduced protein has slightly higher mobility on non-reducing SDS-PAGE.
Source E. coli
Accession Number NM_008371
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Formulation Sterile liquid; 0.1M glycine, pH 3.0, 0.22μm filtered.
Biological Activity The ED50 of this protein, as measured by 2E8 cell proliferation assay, is less than or equal to 1.6ng/mL. This corresponds to a specific activity of greater than or equal to 6.3 x 105 Units/mg.
For Use With (Application) Cytokine Bioassays
Validated Application FA
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Mouse
Gene Symbol Interleukin-7, IL7
Name IL-7
Recombinant Yes

Mouse IL-7 (also called Lymphopoietin 1, LP-1), a pre-B cell growth factor, is an ∽25 kDa factor produced by bone marrow and thymic stromal cells, and spleen cells. IL-7 is a stromal cell-derived growth factor for progenitor B cells and T cells. The main population in the thymus responsive to IL-7 is CD4-CD8-. It also stimulates proliferation and differentiation of mature T cells and NK cells.