Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-6, Recombinant Protein

Mouse Recombinant Protein

$288.55 - $752.55

Specifications

Structural Form E. coli expressed amino acids Phe 25-Thre 211 of mature mouse IL-6 accession # MMINTL6
Purity >98%
Purification Method SDS-PAGE
Molecular Weight The protein is methionylated at the N-terminal. The DTT reduced and non-reduced protein migrate as 19kDa polypeptides on SDS-PAGE.
Source E. coli
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501128715

 
ebioscience™
14806162
5μg Each for $288.55

508953

 
ebioscience™
14806180
25μg Each for $752.55
Description & Specifications

Specifications

Structural Form E. coli expressed amino acids Phe 25-Thre 211 of mature mouse IL-6 accession # MMINTL6
Purity >98%
Purification Method SDS-PAGE
Molecular Weight The protein is methionylated at the N-terminal. The DTT reduced and non-reduced protein migrate as 19kDa polypeptides on SDS-PAGE.
Source E. coli
Accession Number MMINTL6
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Formulation Sterile liquid: 0.1M glycine, pH 3.0, 0.5% BSA. 0.22μm filtered.
Biological Activity The ED50 of this protein, as measured by B9 cell proliferation assay, is less than or equal to 5pg/mL. This corresponds to a specific activity of greater than or equal to 2 x 108 Units/mg.
For Use With (Application) Cytokine Bioassays, ELISA
Validated Application ELISA, FA
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Mouse
Gene Symbol Interleukin-6, IL6
Name IL-6
Recombinant Yes

Mouse IL-6 (also known as B Cell Stimulating Factor, BCSF, and BSF-2) is a ∽21-28 kDa factor produced by a variety of cell types, including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, and glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences the antigen-specific immune response, inflammatory response, and the development of neurons, and is a major mediator of the acute phase reaction.