Please login to your online account to display your discounted pricing

Mouse IL-5 Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gene ID (Entrez) 16191
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

SM5000

 
r & d systems
RDSSM5000
Six 96-well plates 1 Pk. Each for $1,967.07

M5000

 
r & d systems
RDSM5000
two 96-well plates 1 Kit Each for $819.94
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gene ID (Entrez) 16191
Species Mouse

Interleukin 5 (IL-5) is a T cell-derived factor that promotes the proliferation, differentiation and activation of eosinophils. In mice, IL-5 is also a growth and differentiation factor for B cells. Various names previously used to describe IL-5 include: Tcell replacing factor (TRF), B cell growth factor II (BCGFII), B cell differentiation factor μ (BCDF μ), eosinophil differentiation factor (EDF) and eosinophil colony-stimulating factor (Eo-CSF). Biologically active IL-5 is a disulfide-linked homodimer. Rat IL-5 is 94% and 70% identical to mouse and human IL-5, respectively.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 7pg/mL
Specificity: Natural and recombinant mouse IL-5
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.