Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse IL-5 ELISA Kits


Measure levels of mouse interleukin-5 in mouse serum and culture supernatant samples using 96-well plates.

Manufacturer: thermo scientific™ pierce™  PIEMIL5

 View more versions of this product

Catalog No. ENEMIL5

  • / Each
 

Description & Specifications

Specifications

Sensitivity <5pg/mL
Precision Intra-assay CV <10%; inter-assay CV <10%
Sufficient For 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Kit Contents 30X Wash Buffer, 50mL
Formulation Complete ELISA kit with coated 96-well plate
Sample Type Cell culture supernatant, serum
Sample Size 50µL
Total Assay Time 3.5 hr.
Assay Range 20 to 320pg/mL
Specificity No significant cross-reaction with with mouse IL-2, IL-3, IL-4, IL-6, IFNg, GM-CSF, TNFa, rat IL-5, rabbit IL-5 or human IL-5
Target Natural and recombinant mouse interleukin-5 (IL-5)
Quantity 1 plate kit
Description Mouse IL-5 ELISA Kit
Measurement Absorbance at 450nm minus absorbance at 550nm

Assay IL-5 (mouse interleukin-5) levels in mouse serum and culture supernatant samples with our Thermo Scientific™ Mouse IL-5 ELISA Kits.

These mouse IL-5 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-5 (IL-5) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The antibody-pair kit consists of the matched IL-5 antibody pairs and IL-5 standard only. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant mouse interleukin-5 (IL-5)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 20 to 320pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with with mouse IL-2, IL-3, IL-4, IL-6, IFNγ, GM-CSF, TNFα, rat IL-5, rabbit IL-5 or human IL-5

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).