missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-4 Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 16189
Sample Type Cell Culture Supernates, Serum
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
M4000B R & D Systems
RDSM4000B
2 Plates Sufficient materials to run ELISAs on at least two 96-well plates Each for $819.94
SM4000B R & D Systems
RDSSM4000B
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $1,967.07
Specifications

Description

Interleukin 4 (IL-4) is a pleiotropic cytokine produced by activated T cells, mast cells, and basophils. It was initially identified as a B cell differentiation factor (BCDF), as well as a B cell stimulatory factor (BSF1). IL-4 has since been shown to have multiple biological effects on hematopoietic and non-hematopoietic cells, including B and T cells, monocytes, macrophages, mast cells, myeloid and erythroid progenitors, fibroblasts, and endothelial cells. Rat, mouse and human IL-4 are species-specific in their activities.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 2pg/mL
Specificity: Natural and recombinant mouse IL-4
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
16189
Cell Culture Supernates (50μL), Serum (50μL)
Mouse
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum
2pg/mL
Natural and recombinant mouse IL-4