Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-4 ELISPOT Ready-SET-Go!™

Contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-4-secreting cells. This ELISPOT reagent set is pre-titrated for optimal spot development.

$361.05 - $862.75

Specifications

For Use With (Application) ELISPOT
Synonym Interleukin-4
Target IL-4
Storage Requirements 2° to 8°C
Species Mouse
View More Specs
Includes:  Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017444

 
life technologies
LS88784421
2 x 96 tests Each for $361.05

5017445

 
life technologies
LS88784488
10 x 96 tests Each for $862.75
Description & Specifications

Specifications

For Use With (Application) ELISPOT
Synonym Interleukin-4
Target IL-4
Storage Requirements 2° to 8°C
Species Mouse