missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-33 DuoSet, R&D Systems

$846.01 - $846.01

Specifications

Gene ID (Entrez) 77125
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DY3626 R & D Systems
RDSDY3626
Each for $846.01
Specifications

Description

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

77125