missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse IL-31 Platinum ELISA Kit

For the quantitative detection of mouse IL-31.

Manufacturer:  Invitrogen™ LSBMS6030TEN

 View more versions of this product

Catalog No. 501125449



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-31
Biotin-Conjugate anti-mouse IL-31 monoclonal antibody
Streptavidin-HRP
Mouse IL-31 Standard lyophilized, 2000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
9.1pg/mL
50μL
IL-31
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Mouse
Interleukin-31
Documents