Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-27 ELISA Ready-SET-Go!™ Kit

Engineered for accurate and precise measurement of mouse IL-27 protein levels from samples including serum, and cell culture supernatants.This ELISA has 11.8% cross reactivity to the recombinant human IL-27 and no cross reactivity to the recombinant mouse IL-12 when spiked in excess (100ng/ml)

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym Interleukin-27
Target IL-27
Sensitivity 16pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017305

 
life technologies
LS88727422
Each for $332.05

5017306

 
life technologies
LS88727486
96-well plates 10 x 96 tests Each for $694.55

5017307

 
life technologies
LS88727488
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-27
Target IL-27
Sensitivity 16pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Mouse

Mouse IL-27 Ready-SET-Go! ELISA Set (with or without high affinity binding ELISA microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)

  • To ensure optimal results from this ELISA Ready-SET-Go! Set, please only use the components included in the set. Exchanging of components may lead to a change in signal