Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-2 ELISPOT Ready-SET-Go!™

Contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IL-2-secreting cells. This ELISPOT reagent set is pre-titrated for optimal spot development.

$361.05 - $862.75

Specifications

For Use With (Application) ELISPOT
Synonym Interleukin-2
Target IL-2
Storage Requirements 2° to 8°C
Species Mouse
View More Specs
Includes:  Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer Powder.Reconstitute to 1L with dH20 and filter (0.22 uM)).
5X ELISA/ELISPOT Diluent.
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017436

 
life technologies
LS88782421
2 x 96 tests Each for $361.05

5017437

 
life technologies
LS88782488
10 x 96 tests Each for $862.75
Description & Specifications

Specifications

For Use With (Application) ELISPOT
Synonym Interleukin-2
Target IL-2
Storage Requirements 2° to 8°C
Species Mouse