missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-2 DuoSet, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Mouse IL-2

$324.58 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Quantity 1 Kit
Gene ID (Entrez) 16183
Species Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DY402 R & D Systems
RDSDY402
15 Plates Each for $846.01
DY40205 R & D Systems
RDSDY40205
5 Plates Each for $324.58
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit
Mouse
ELISA Development Kit
16183