Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse IL-1beta Platinum ELISA Kit

For the quantitative detection of mouse IL-1beta.

$651.05 - $3,116.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Catabolin, Interleukin-1 beta, IL1F2
Target IL-1 beta
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-1beta
Biotin-Conjugate anti-mouse IL-1beta monoclonal antibody
Streptavidin-HRP
Mouse IL-1beta Standard lyophilized, 1ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018265

 
ebioscience&trade
LSBMS6002
96 tests Each for $651.05

5018267

 
ebioscience&trade
LSBMS6002TEN
10 x 96 tests Each for $3,116.05

5018268

 
ebioscience&trade
LSBMS6002TWO
2 x 96 tests Each for $854.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Catabolin, Interleukin-1 beta, IL1F2
Target IL-1 beta
Conjugate Biotin
Sample Type Cell culture supernatant, serum
Sensitivity 1.2pg/mL
Species Mouse

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.

IL-1beta is a pro-inflamatory cytokine and is expressed by many cells including macrophage, NK cells, monocytes, and neutrophils.