Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse IL-17F Platinum ELISA Kit

For the quantitative detection of mouse IL-17F.

$651.05 - $3,116.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin 17F, Cytokine ML1
Target IL-17F
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-17F
Biotin-Conjugate anti-mouse IL-17F polyclonal antibody
Streptavidin-HRP
Mouse IL-17F Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Conjugate Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Calibrator Diluent
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125443

 
ebioscience
LSBMS6020
96 tests Each for $651.05

5018298

 
ebioscience&trade
LSBMS6020TEN
10 x 96 tests Each for $3,116.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin 17F, Cytokine ML1
Target IL-17F
Conjugate Biotin
Sample Type Cell culture supernatant, serum and plasma (EDTA)
Sensitivity 10pg/mL
Species Mouse

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.