Please login to your online account to display your discounted pricing

eBioscience Mouse IL-17F (homodimer) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of mouse IL-17FF homodimer protein levels from samples including serum, and cell culture supernatants.

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym Interleukin-17F, IL-17FF
Target IL-17F
Sensitivity 15pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

 Alert

Contains Azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125206

 
ebioscience
88747286
96-well plates 10 x 96 tests Each for $694.55

5017400

 
ebioscience
88747222
Each for $332.05

5017402

 
ebioscience
88747288
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-17F, IL-17FF
Target IL-17F
Sensitivity 15pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Mouse

Mouse IL-17F ELISA Ready-SET-Go! Reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)

  • The assay demonstrates parallelism in measuring recombinant and native mouse IL-17F proteins and a standard curve range of 15 to 2000pg/mL
  • No crossreactivity to the IL-17AA homodimer is observed and <2.5% crossreactivity to IL-17AF heterodimer when spiked in excess (100ng/ml)