Please login to your online account to display your discounted pricing

eBioscience™ Mouse IL-17E (IL-25) ELISA Ready-SET-Go!™ Kit

Optimized for the accurate and precise measurement of mouse IL-17E in serum and tissue culture supernatant samples

$332.05 - $565.72

Specifications

For Use With (Application) ELISA
Detects IL-17E (IL-25)
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Sample Diluent B
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017175

 
life technologies
LS88700222
2 x 96 tests Each for $332.05

5017177

 
life technologies
LS88700288
10 x 96 tests Each for $565.72
Description & Specifications

Specifications

For Use With (Application) ELISA
Detects IL-17E (IL-25)
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C

  • Mouse IL-17E ELISA Ready-SET-Go! reagent set contains all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays
  • Serum samples must be diluted 8-fold in the provided sample diluent prior to evaluation in this assay