missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ IL-17AF (heterodimer) Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.
$380.00 - $924.00
Specifications
Target Species | Mouse |
---|---|
For Use With (Application) | ELISA |
Synonym | Interleukin-17A/F |
Alert:
Contains Sodium azide
Catalog Number | Mfr. No. | Quantity | Gene Symbol | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Gene Symbol | Price | Quantity | ||||
501125230
|
Invitrogen™
88871188 |
10 x 96 Tests | GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 |
Each for $924.00
|
|
||||
501125226
|
Invitrogen™
88871122 |
2 x 96 Tests | Ctla-8,Ctla8,IL-17,IL-17A,Il17 |
Each for $380.00
|
|
||||
Description
- Mouse IL-17AF (heterodimer) ELISA kit contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
- By incorporating a monoclonal antibody to IL-17A as the capture reagent and IL-17AF as the detection reagent, this second generation kit has increased sensitivity with a range from 16 to 2000pg/mL and no cross-reactivity to the IL-17A or IL-17F homodimers
Specifications
Mouse | |
ELISA | |
Interleukin-17A/F |
For Research Use Only