missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse IL-17AF (heterodimer) ELISA Ready-SET-Go!™ Kit, 2nd Generation

Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.

Manufacturer:  Invitrogen™ LS88871186

 View more versions of this product

Catalog No. 501125229


Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer 10X PBS ELISA Coating Buffer.
Assay Diluent. 5X concentrated
Sample Diluent A
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

  • Mouse IL-17AF (heterodimer) Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
  • By incorporating a monoclonal antibody to IL-17A as the capture reagent and IL-17AF as the detection reagent, this second generation kit has increased sensitivity with a range from 16 to 2000pg/mL and no cross-reactivity to the IL-17A or IL-17F homodimers
Specifications

Specifications

ELISA
10 x 96 tests
Mouse
IL-17AF
96-well plates
16pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-17A/F
Documents

For Research Use Only