missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse IL-13 Platinum ELISA Kit

For the quantitative detection of mouse IL-13.

Manufacturer:  Invitrogen™ LSBMS6015FIVE

 View more versions of this product

Catalog No. 501125442



Includes: Capture Antibody Concentrate (250 x)
Detection Antibody Concentrate (250 x)
mouse IL-19 standard, (lyophilized), 8000pg/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS) 10x
Assay Buffer A Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Tetramethylbenzidine (TMB) Substrate Solution
2 96-well plates

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
5 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Mouse
ALRH, BHR1, allergic rhinitis
ELISA
2.8pg/mL
50μL
IL-13
Documents

For Research Use Only