Please login to your online account to display your discounted pricing

Mouse IL-13 DuoSet, R&D Systems

$317.73 - $828.14

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Gene ID (Entrez) 16163
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DY413

 
r & d systems
RDSDY413
15 96-well plates Each for $828.14

DY41305

 
r & d systems
DY41305
5 Plate Each for $317.73
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Gene ID (Entrez) 16163
Species Mouse

Assay Type: ELISA Development Kit
Sensitivity: 0pg/mL
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.