missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-13 Culture Media Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Gene ID (Entrez) 16163
Sample Type Cell Culture Supernates
Sample Volume Cell Culture Supernates (50μL)
Sensitivity 1.5pg/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1300CB R & D Systems
RDSM1300CB
2 Plates Each for $819.94
SM1300CB R & D Systems
RDSSM1300CB
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL)
Sensitivity: 1.5pg/mL
Specificity: Natural and recombinant mouse IL-13
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Cell Culture Supernates
1.5pg/mL
Natural and recombinant mouse IL-13
16163
Cell Culture Supernates (50μL)
Mouse