missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-12 (total) ELISA Kits, Thermo Scientific™

Microplate kits for sandwich ELISA of total mouse interleukin-12 (IL-12, p70 plus p40)

$2,436.00 - $2,436.00


Includes: Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
EMIL12TOT5 Pierce Biotechnology Inc
EMIL12TOT5
Each for $2,436.00
Specifications

Description

These mouse IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of both active (p70) and inactive (p40) forms of interleukin-12 (IL-12 total) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Total (p70 and p40) natural and recombinant mouse interleukin-12 (IL-12)
  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
  • Measurement: Absorbance at 450nm minus absorbance at 550nm
  • Assay Range: 47 to 3000pg/mL
  • Sensitivity: < 12pg/mL
  • Precision: Dependent upon sample type (generally, intra- and inter-assay CV < 10%)
  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
  • Total Assay Time: 3 hours
  • Sample Size: 50μL
  • Specificity: No significant cross-reaction with mouse G-CSF, GM-CSF, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MCP-1, MIP-1α, RANTES or TNFα, human IL-12, or pig IL-12