missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-12 p70 Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 16159
Sample Type Cell Culture Supernates, Serum
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1270 R & D Systems
RDSM1270
2 Plates Each for $819.94
SM1270 R & D Systems
RDSSM1270
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 2.5pg/mL
Specificity: Natural and recombinant mouse IL-12 p70
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16159
Cell Culture Supernates (50μL), Serum (50μL)
Mouse
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum
2.5pg/mL
Natural and recombinant mouse IL-12 p70