missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse IL-12 (p40) ELISA Kits

Measure levels of inactive mouse interleukin-12, p40 in mouse serum, plasma and culture supernatant samples using 96-well plates.

$651.05 - $1,145.50

Specifications

Assay Range 12.5 to 800pg/mL
Description Mouse IL-12 (p40) ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
ENEMIL12P40 Thermo Scientific™ Pierce™
PIEMIL12P40
96 microplate wells Each for $651.05
EEMIL12P402 Thermo Scientific™ Pierce™
EMIL12P402
2 x 96 microplate wells Each for $1,145.50
Specifications

Description

Assay IL-12 p40 (inactive mouse interleukin-12, p40) levels in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Mouse IL-12 (p40) ELISA Kits.

These mouse IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of biologically inactive interleukin-12 (IL-12 p40) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interleukin-12 p40 (IL-12 p40)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 12.5 to 800pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-18, Eotaxin, MCP-1, MIP-1α, RANTES or TNFα or human IL-12 p40. Mouse IL-12 p70 exhibits 1% cross-reactivity in this assay

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Description & Specifications

Specifications

12.5 to 800pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with mouse IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-18, Eotaxin, MCP-1, MIP-1a, RANTES or TNFa or human IL-12 p40. Mouse IL-12 p70 exhibits 1% cross-reactivity in this assay
3 hr.
Mouse IL-12 (p40) ELISA Kit
Absorbance at 450nm minus absorbance at 550nm
50µL
<3pg/mL
Natural and recombinant mouse interleukin-12 p40 (IL-12 p40)