missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-12/IL-23 p40 NonAllele-specific Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Gene ID (Entrez) 16160
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Sensitivity 2.7pg/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MP400 R & D Systems
RDSMP400
2 Plates Each for $819.94
SMP400 R & D Systems
RDSSMP400
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Sensitivity: 2.7pg/mL
Specificity: Natural and recombinant mouse IL-12/IL-23 p40
Cross Reactivity: <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
2.7pg/mL
Natural and recombinant mouse IL-12/IL-23 p40
16160
Cell Culture Supernates (50μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Mouse