Please login to your online account to display your discounted pricing

Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet, R&D Systems

$317.73 - $828.14

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Quantity 1 Kit
Gene ID (Entrez) 16160
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DY2398

 
r & d systems
RDSDY2398
15 Plates Each for $828.14

DY239805

 
r & d systems
RDSDY239805
5 Plates Each for $317.73
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Quantity 1 Kit
Gene ID (Entrez) 16160
Species Mouse

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.