missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-12/IL-23 p40 Allele-specific DuoSet, R&D Systems

$324.58 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Quantity 1 Kit
Gene ID (Entrez) 16160
Species Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DY499 R & D Systems
RDSDY499
15 Plates Each for $846.01
DY49905 R & D Systems
RDSDY49905
5 Plates Each for $324.58
Specifications

Description

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit
Mouse
ELISA Development Kit
16160