missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IL-10 Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 16153
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (25μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1000B R & D Systems
RDSM1000B
2 Plates Each for $819.94
SM1000B R & D Systems
RDSSM1000B
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (25μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Sensitivity: 5.22pg/mL
Specificity: Natural and recombinant mouse IL-10
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16153
Cell Culture Supernates (25μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Mouse
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
5.22pg/mL
Natural and recombinant mouse IL-10