Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse IL-10 Platinum ELISA Kit

For the quantitative detection of mouse IL-10.

$651.05 - $3,116.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym TGIF, MCSF III, CSIF, B TCGF
Target IL-10
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-10
Biotin-Conjugate anti-mouse IL-10 polyclonal antibody
Streptavidin-HRP
Mouse IL-10 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125461

 
ebioscience
LSBMS6142FIVE
5 x 96 tests Each for $1,869.05

5018327

 
ebioscience&trade
LSBMS6142
96 tests Each for $651.05

5018329

 
ebioscience&trade
LSBMS6142TEN
10 x 96 tests Each for $3,116.05

5018330

 
ebioscience&trade
LSBMS6142TWO
2 x 96 tests Each for $854.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym TGIF, MCSF III, CSIF, B TCGF
Target IL-10
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate)
Sensitivity 5.0pg/mL
Species Mouse

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency