missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Mouse IL-10 ELISA Ready-SET-Go!™ Kit, 2nd Generation
Specifically engineered for accurate and precise measurement of mouse IL-10 protein levels from samples including serum and cell culture supernatants. This 2nd Generation kit has decreased background and improved performance in serum samples.
Manufacturer: Invitrogen™ LS88710586
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer 10X PBS ELISA Coating Buffer.
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution
Description
Interleukin-10 is a pleiotropic cytokine playing an important role as a regulator of lymphoid and myeloid cell function. In humans IL-10 is produced by activated CD8(+) peripheral blood T-cells, by T-helper CD4(+) T-cell clones. Due to the ability of IL-10 to block cytokine synthesis and several accessory cell functions of macrophages this cytokine is a potent suppressor of the effector functions of macrophages, T-cells and NK cells. In addition, IL-10 participates in regulating proliferation and differentiation of B-cells, mast cells and thymocytes. The primary structure of human IL-10 has been determined by cloning the cDNA encoding the cytokine. The corresponding protein exerts 160 amino acids with a predicted molecular mass of 18.5 kDa. Based on its primary structure, IL-10 is a member of the four a-helix bundle family of cytokines. In solution human IL-10 is a homodimer with an apparent molecular mass of 39 kDa. Although it contains an N-linked, glycosylation site, it lacks detectable carbohydrates. Recombinant protein expressed in E. coli thus retains all known biological activities. The human IL-10 gene is located on chromosome 1 and is present as a single copy in the genome. The human IL-10 exhibits strong DNA and amino acid sequence homology to the murine IL-10 and an open reading frame in the Epstein-Barr virus genome, BCRF1 which shares many of the cellular cytokine's biological activities and may therefore play a role in the host-virus interaction. The immunosuppressive properties of IL-10 suggest a possible clinical use of IL-10 in suppressing rejections of grafts after organ transplantations. IL-10 can furthermore exert strong anti-inflammatory activities.Specifications
P18893 | |
Biotin | |
16153 | |
Colorimetric Microplate Reader | |
ELISA | |
RUO | |
Serum, Supernatants | |
Mouse | |
1 hr 45 min | |
Mouse | |
Interleukin 10, CSIF, Cytokine Synthesis Inhibitory Factor |
32-4,000 pg/mL | |
ELISA | |
96-well plates | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, Coating Buffer 10X PBS ELISA Coating Buffer., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86) | |
10 x 96 tests | |
32 pg/mL | |
Serum (100μL), Supernatants (100μL) | |
2-8°C | |
24 hr 30 min | |
Il10 | |
HRP |
For Research Use Only