Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse IL-1 beta ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of mouse IL-1 beta protein levels from samples including serum, protein lysate and cell culture supernatants. This ELISA set will recognize the cleaved, mature form and uncleaved pro-form of Mouse IL-1 beta.

$303.05 - $999.05

Specifications

For Use With (Application) ELISA
Synonym Interleukin-1 beta
Target IL-1 beta
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017185

 
ebioscience&trade
LS88701377
Each for $941.05

501128706

 
ebioscience
LS88701376
96-well plates 20 x 96 tests Each for $999.05

501128807

 
ebioscience
LS88701386
96-well plates 10 x 96 tests Each for $694.55

501128814

 
ebioscience
LS88701388
Each for $665.55

501129749

 
ebioscience&trade
LS88701322
Each for $303.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-1 beta
Target IL-1 beta
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C
Species Mouse

Mouse IL-1 beta ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)

  • To ensure optimal results from the Mouse IL-1 beta ELISA Ready-SET-Go! Set, please only use the components included in the set. Exchanging of components may lead to a change in signal