Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse IL-1 beta ELISA Kits


Measure levels of mouse interleukin-1 beta (IL-1b) in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

Manufacturer: thermo scientific™ pierce™  EM2IL1B

 View more versions of this product

Catalog No. ENEM2IL1B

  • / Each
 

Description & Specifications

Specifications

Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sensitivity <3pg/mL
Kit Contents 20X Wash Buffer, 55mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 1 plate kit
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Description Mouse IL-1 beta ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Formulation Complete ELISA kit with coated 96-well plate
Assay Range 31.2 to 2000pg/mL
Sample Size 50µL
Specificity No significant cross-reaction with mouse IL-1a, IL-10, IL-2, IL-4, IL-6, TNFa, GM-CSF; human IL-1ß, IL-8, IL-1a; or bovine IL-1ß. This ELISA does cross-react with rat IL-1ß (12%)
Sufficient For 96 microplate wells
Total Assay Time 4 hr.
Target Natural and recombinant mouse interleukin-1 beta (IL-1ß)

Assay IL-1β (mouse interleukin-1 beta) levels in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Mouse IL-1 beta ELISA Kits.

These mouse IL-1 beta ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-1 beta (IL-1β) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant mouse interleukin-1 beta (IL-1β)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 31.2 to 2000pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-1α, IL-10, IL-2, IL-4, IL-6, TNFα, GM-CSF; human IL-1β, IL-8, IL-1α; or bovine IL-1β. This ELISA does cross-react with rat IL-1β (12%)

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.